Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCDFP-15 Monoclonal Antibody (23A3)
GREENER_CHOICE

Catalog No. PIMA511633
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA511633 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA511633 Supplier Invitrogen™ Supplier No. MA511633
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-11633 targets GCDFP-15 in IHC (P) applications and shows reactivity with Human and Rat samples. The MA5-11633 immunogen is recombinant protein encoding the excreted domain of human GCDFP15.

Gross cystic disease of the breast is benign premenopausal disorder in which cysts are a predominant pathological lesion. These cysts appear to be formed from excessive apocrine cystic secretions. This fluid is composed of several glycoproteins including a unique 15kDa monomer protein, Gross Cystic Disease Fluid Protein-15 (GCDFP15). Cytosolic analysis of normal tissue specimens from all major organs has demonstrated GCDFP15 in apocrine epithelia, lacrimal, ceruminous and Moll's glands and in numerous serous cells of the submandibular, tracheal, bronchial, sublingual and minor salivary glands. GCDFP15 and prostate specific antigen are co-expressed in androgen receptor-positive breast tumours.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCDFP-15
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 23A3
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation tissue culture supernatant with 0.09% sodium azide
Gene PIP
Gene Accession No. O70417, P12273
Gene Alias 14 kDa submandibular gland protein; gcdfp 15; GCDFP15; GCDFP-15; gp17; GPIP4; Gross cystic disease fluid protein 15; HGNC:8993; mSMGP; PIP; prolactin induced protein; prolactin-induced protein; Prolactin-inducible protein; prolactin-inducible protein homolog; SABP; secretory actin-binding protein; SMGP
Gene Symbols PIP
Host Species Mouse
Immunogen Recombinant protein encoding the excreted domain of human GCDFP15.
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5304, 64673
Target Species Human, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.