Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCDFP-15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582381
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582381 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582381 Supplier Invitrogen™ Supplier No. PA582381
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FDIPKSVRPN DEVTAVLAVQ TELKECMVVK TYLISSIPLQ GAFNYKYTAC LCDDNPKTFY WDFYTNRTVQ IAAVVDVIRE LGICPDDAAV IPIKNNRFYT IEILKVE Highest antigen sequence indentity to the following orthologs: Rat - 43%, Mouse - 47%.

Gross cystic disease of the breast is benign premenopausal disorder in which cysts are a predominant pathological lesion. These cysts appear to be formed from excessive apocrine cystic secretions. This fluid is composed of several glycoproteins including a unique 15kDa monomer protein, Gross Cystic Disease Fluid Protein-15 (GCDFP15). Cytosolic analysis of normal tissue specimens from all major organs has demonstrated GCDFP15 in apocrine epithelia, lacrimal, ceruminous and Moll's glands and in numerous serous cells of the submandibular, tracheal, bronchial, sublingual and minor salivary glands. GCDFP15 and prostate specific antigen are co-expressed in androgen receptor-positive breast tumours.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCDFP-15
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIP
Gene Accession No. P12273
Gene Alias 14 kDa submandibular gland protein; gcdfp 15; GCDFP15; GCDFP-15; gp17; GPIP4; Gross cystic disease fluid protein 15; HGNC:8993; mSMGP; PIP; prolactin induced protein; prolactin-induced protein; Prolactin-inducible protein; prolactin-inducible protein homolog; SABP; secretory actin-binding protein; SMGP
Gene Symbols PIP
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIP. Recombinant protein control fragment (Product #RP-89946).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5304
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.