Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCET2 Antibody (HGAL/2373), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314265AF594
This item is not returnable.
View return policy
Description
GCET2 Monoclonal antibody specifically detects GCET2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
GCET2 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
HGAL/2373 | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
GAL, GCAT2, germinal center B-cell-expressed transcript 2 protein, germinal center expressed transcript 2, Germinal center-associated lymphoma protein, HGAL, MGC40441 | |
Recombinant fragment (178 Amino acid residues from around aa1-200) of human GCET2 protein (Uniprot: Q8N6F7) | |
0.1 mL | |
Cancer, Immunology | |
257144 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction