Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCFC1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171923AF700
Description
GCFC1 Polyclonal antibody specifically detects GCFC1 in Human samples. It is validated for Western BlotSpecifications
GCFC1 | |
Polyclonal | |
Western Blot | |
BM020, C21orf66, chromosome 21 open reading frame 66, FLJ90561, fSAP105, functional spliceosome-associated protein 105, GCFCFSAP105, GC-rich sequence DNA-binding factor 1, GC-rich sequence DNA-binding factor candidate | |
The immunogen for this product maps to a region between residue 867 and 917 of human GC-rich Sequence DNA-binding Factor 1 using the numbering given in entry NP_057715.2 (GeneID 94104). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
94104 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction