Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCFC1 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299179F
Description
GCFC1 Polyclonal antibody specifically detects GCFC1 in Human samples. It is validated for Western BlotSpecifications
GCFC1 | |
Polyclonal | |
Western Blot | |
BM020, C21orf66, chromosome 21 open reading frame 66, FLJ90561, fSAP105, functional spliceosome-associated protein 105, GCFCFSAP105, GC-rich sequence DNA-binding factor 1, GC-rich sequence DNA-binding factor candidate | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GCFC1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
FITC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
94104 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction