Learn More
GCFC1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303166A

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that may bind to GC-rich DNA sequences, which suggests its involvement in the regulation of transcription. Alternative splicing of this gene results in multiple transcript variants.Specifications
GCFC1 | |
Polyclonal | |
Unconjugated | |
PAXBP1 | |
BM020, C21orf66, chromosome 21 open reading frame 66, fSAP105, functional spliceosome-associated protein 105, GCFC, GCFC1, GC-rich sequence DNA-binding factor 1, GC-rich sequence DNA-binding factor candidate | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
94104 | |
4° C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9Y5B6 | |
PAXBP1 | |
Between 225 and 275 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.