Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCFC2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP105985
Description
GCFC2 Polyclonal specifically detects GCFC2 in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).Specifications
GCFC2 | |
Polyclonal | |
Unconjugated | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
C2orf3, chromosome 2 open reading frame 3, DNABF, GC bindng factor, GCF, GCFGC binding factor, GC-rich sequence DNA-binding factor, GC-rich sequence DNA-binding factor 2, TCF9, TCF-9, Transcription factor 9, transcription factor 9 (binds GC-rich sequences) | |
Rabbit | |
Affinity Purified | |
RUO | |
6936 | |
Store at 4C. Do not freeze. |
Immunoprecipitation | |
1.0 mg/mL | |
Immunoprecipitation 1:10-1:500 | |
NP_003194.3 | |
GCFC2 | |
The immunogen recognized by this antibody maps to a region between residue 731 and 781 of human GC-rich sequence DNA-binding factor using the numbering given in entry NP_003194.3 (GeneID 6936). | |
100 μL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction