Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517711
Description
It is not recommended to aliquot this antibody. This antibody is not cross-reactive with HPK or other members of the GCK family.
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Alternative splicing of this gene results in three tissue-specific forms of glucokinase, one found in pancreatic islet beta cells and two found in liver. The protein localizes to the outer membrane of mitochondria. In contrast to other forms of hexokinase, this enzyme is not inhibited by its product glucose-6-phosphate but remains active while glucose is abundant. Mutations in this gene have been associated with non-insulin dependent diabetes mellitus (NIDDM), maturity onset diabetes of the young, type 2 (MODY2) and persistent hyperinsulinemic hypoglycemia of infancy (PHHI).
Specifications
GCK | |
Polyclonal | |
0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
P17712, P35557, P52792 | |
GCK | |
Synthetic peptide corresponding to residues surrounding Ser380 of human GCK. | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
GCK | |
ATP:D-hexose 6-phosphotransferase; FGQTL3; GC kinase; Gck; Gk; GLK; Gls006; glucokinase; glucokinase (hexokinase 4); glucokinase variant 3; GLUKA; hexokinase 4; hexokinase D, pancreatic isozyme; hexokinase type IV; Hexokinase-4; hexokinase-D; HHF3; HK IV; HK4; HKIV; Hlb62; HXKP; LGLK; MEKKK 2; MODY2; RNGK2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
103988, 24385, 2645 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction