Learn More
Invitrogen™ GCK Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA590199
Description
Immunogen sequence: RMCVNTEWGA FGDSGELDEF LLEYDRLVDE SSANPGQQLY EKLIGGKYMG ELVRLVLLRL VDENLLFHGE ASEQLRTRGA FETRFVSQVE SDTGDRKQIY NILSTLGLRP STTDCDIVRR ACESVSTRAA HMCSAGLAGV INRMRESRSE DVMRITVGVD GSVYKLHPSF KERFHASVRR LTPSCEITFI ESEEGSGRGA ALVSAVACKK ACMLGQ; Positive Samples: LO2, A-549, mouse liver, rat liver; Cellular Location: Cytoplasm, Nucleus.
Hexokinases phosphorylate glucose to produce glucose-6-phosphate, thus committing glucose to the glycolytic pathway. Alternative splicing of this gene results in three tissue-specific forms of glucokinase, one found in pancreatic islet beta cells and two found in liver. The protein localizes to the outer membrane of mitochondria. In contrast to other forms of hexokinase, this enzyme is not inhibited by its product glucose-6-phosphate but remains active while glucose is abundant. Mutations in this gene have been associated with non-insulin dependent diabetes mellitus (NIDDM), maturity onset diabetes of the young, type 2 (MODY2) and persistent hyperinsulinemic hypoglycemia of infancy (PHHI).
Specifications
| GCK | |
| Polyclonal | |
| Unconjugated | |
| GCK | |
| ATP:D-hexose 6-phosphotransferase; FGQTL3; GC kinase; Gck; Gk; GLK; Gls006; glucokinase; glucokinase (hexokinase 4); glucokinase variant 3; GLUKA; hexokinase 4; hexokinase D, pancreatic isozyme; hexokinase type IV; Hexokinase-4; hexokinase-D; HHF3; HK IV; HK4; HKIV; Hlb62; HXKP; LGLK; MEKKK 2; MODY2; RNGK2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 103988, 24385, 2645 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.55 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P17712, P35557, P52792 | |
| GCK | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.