Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCKR Antibody, Novus Biologicals™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP128910
Description
GCKR Polyclonal specifically detects GCKR in Mouse, Rat samples. It is validated for Western Blot, Peptide ELISA.Specifications
GCKR | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
FGQTL5, GKRP, glucokinase (hexokinase 4) regulator, glucokinase (hexokinase 4) regulatory protein, Glucokinase regulator, glucokinase regulatory protein | |
Goat | |
69 kDa | |
0.1 mg | |
Primary | |
Cross reacts with Mouse and Rat. Expected from sequence similarity: Human. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:8000 | |
Q14397 | |
GCKR | |
Peptide with sequence KRFQHVIETPEPGK -C corresponding to N-Terminus according to NP_001477.1. | |
Affinity Purified | |
RUO | |
2646 | |
Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction