Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCKR Monoclonal Antibody (OTI9G7), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502163S


Description
GCKR Monoclonal antibody specifically detects GCKR in Human samples. It is validated for Immunocytochemistry, Western BlotSpecifications
GCKR | |
Monoclonal | |
0.6 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q96IQ7 | |
Mouse | |
Affinity chromatography | |
RUO | |
23584 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG1 |
Immunocytochemistry, Western Blot | |
OTI9G7 | |
Unconjugated | |
VSIG2 | |
FGQTL5; Gckr; GKRP; GLRE; glucokinase (hexokinase 4) regulator; glucokinase regulator; Glucokinase regulatory protein; hexokinase 4 | |
Full length human recombinant protein of human VSIG2 produced in HEK293T cell. | |
30 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction