Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCKR Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA527959
Description
Recommended positive controls: 293T, A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse liver. Predicted reactivity: Mouse (89%), Rat (90%), Dog (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein belonging to the GCKR subfamily of the SIS (Sugar ISomerase) family of proteins. The gene product is a regulatory protein that inhibits glucokinase in liver and pancreatic islet cells by binding non-covalently to form an inactive complex with the enzyme. This gene is considered a susceptibility gene candidate for a form of maturity-onset diabetes of the young (MODY).
Specifications
| GCKR | |
| Polyclonal | |
| Unconjugated | |
| Gckr | |
| FGQTL5; Gckr; GKRP; GLRE; glucokinase (hexokinase 4) regulator; glucokinase regulator; Glucokinase regulatory protein; hexokinase 4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 231103, 2646 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q14397, Q91X44 | |
| Gckr | |
| Recombinant protein encompassing a sequence within the center region of human GCKR. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction