Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCLC Recombinant Rabbit Monoclonal Antibody (0X1H3)
GREENER_CHOICE

Catalog No. PIMA535522
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA535522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA535522 Supplier Invitrogen™ Supplier No. MA535522
Add to Cart
Add to Cart

GCLC Recombinant Rabbit Monoclonal Antibody (ARC1065)

GCLC Recombinant Monoclonal Antibody for Western Blot

Glutamate-cysteine ligase, also known as gamma-glutamylcysteine synthetase is the first rate-limiting enzyme of glutathione synthesis. The enzyme consists of two subunits, a heavy catalytic subunit and a light regulatory subunit. This locus encodes the catalytic subunit, while the regulatory subunit is derived from a different gene located on chromosome 1p22-p21. Mutations at this locus have been associated with hemolytic anemia due to deficiency of gamma-glutamylcysteine synthetase and susceptibility to myocardial infarction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCLC
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone 0X1H3
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Gclc
Gene Accession No. P19468
Gene Alias cb1049; D9Wsu168e; EGK_15023; fe36e11; gamma GCS-HS; gamma-ECS; Gamma-glutamylcysteine synthetase; gamma-glutamylcysteine synthetase heavy subunit; GCL; Gclc; GCS; GCS heavy chain; Ggcs-hs; GLCL; GLCLC; GLCL-H; glutamate-cysteine ligase catalytic subunit; Glutamate--cysteine ligase catalytic subunit; glutamate-cysteine ligase, catalytic subunit; Glutamylcysteine gamma synthetase light chain; wu:fe36e11
Gene Symbols Gclc
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 538-637 of human GCLC (P48506).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25283
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.