Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCLC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583330
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583330 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583330 Supplier Invitrogen™ Supplier No. PA583330
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FENSAYVVFV VLLTRVILSY KLDFLIPLSK VDENMKVAQK RDAVLQGMFY FRKDICKGGN AVVDGCGKAQ NSTELAAEEY TLMSIDTIIN GKEGVFPGLI P Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 94%.

Glutamate-cysteine ligase, also known as gamma-glutamylcysteine synthetase is the first rate-limiting enzyme of glutathione synthesis. The enzyme consists of two subunits, a heavy catalytic subunit and a light regulatory subunit. This locus encodes the catalytic subunit, while the regulatory subunit is derived from a different gene located on chromosome 1p22-p21. Mutations at this locus have been associated with hemolytic anemia due to deficiency of gamma-glutamylcysteine synthetase and susceptibility to myocardial infarction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCLC
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gclc
Gene Accession No. P48506
Gene Alias cb1049; D9Wsu168e; EGK_15023; fe36e11; gamma GCS-HS; gamma-ECS; Gamma-glutamylcysteine synthetase; gamma-glutamylcysteine synthetase heavy subunit; GCL; Gclc; GCS; GCS heavy chain; Ggcs-hs; GLCL; GLCLC; GLCL-H; glutamate-cysteine ligase catalytic subunit; Glutamate--cysteine ligase catalytic subunit; glutamate-cysteine ligase, catalytic subunit; Glutamylcysteine gamma synthetase light chain; wu:fe36e11
Gene Symbols Gclc
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GCLC. Recombinant protein control fragment (Product #RP-94586).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2729
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.