Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCLM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529908
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529908 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529908 Supplier Invitrogen™ Supplier No. PA529908
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa, A431, mouse liver, rat liver, rat kidney. Predicted reactivity: Mouse (97%), Rat (96%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Gamma-glutamylcysteine synthetase (gamma-GCS) is the rate limiting enzyme for glutathione (L-gamma-glutamyl-L-cysteinylglycine, GSH) synthesis. GSH is ubiquitous in mammalian cells as a vital intra- and extracellular protective antioxidant. Gamma-GCS is a heterodimer of a heavy catalytic subunit and a light regulatory subunit that is responsive to inflammation, phenolic ntioxidants, heat shock, oxidants and cytokines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCLM
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.21 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Gclm
Gene Accession No. O09172, P48507, P48508
Gene Alias AI649393; gamma glutamylcysteine synthetase; gamma-ECS regulatory subunit; Gamma-glutamylcysteine synthetase regulatory subunit; Gclm; Gcmc; GCS light chain; Glclr; glutamate cysteine ligase (gamma-glutamylcysteine synthetase), regulatory; glutamate cysteine ligase, modifier subunit; glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory (30.8kD); glutamate-cysteine ligase modifier subunit; glutamate--cysteine ligase modifier subunit; glutamate-cysteine ligase modifier subunit delta2 alternative splicing; glutamate-cysteine ligase regulatory protein; Glutamate--cysteine ligase regulatory subunit; glutamate-cysteine ligase, modifier subunit; GSC light chain; null
Gene Symbols Gclm
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GCLM. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14630, 2730, 29739
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.