Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCLM Rabbit anti-Human, Clone: 0U3J2, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31538020UL
Description
GCLM Monoclonal antibody specifically detects GCLM in Human samples. It is validated for Western BlotSpecifications
GCLM | |
Monoclonal | |
Unconjugated | |
Gamma-ECS regulatory subunit, Gamma-glutamylcysteine synthetase regulatory subunit, GCS light chain, GLCLRglutamate--cysteine ligase regulatory subunit, glutamate-cysteine ligase (gamma-glutamylcysteine synthetase), regulatory(30.8kD), Glutamate--cysteine ligase modifier subunit, glutamate-cysteine ligase regulatory protein, glutamate-cysteine ligase, modifier subunit, GSC light chain | |
A synthetic peptide corresponding to a sequence within amino acids 175-274 of human GCLM (P48507). | |
20 μg | |
Cancer | |
2730 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
0U3J2 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction