Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCM2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982794
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2794 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2794 Supplier Bioss Supplier No. BS13314R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene is a homolog of the Drosophila glial cells missing gene, which is thought to act as a binary switch between neuronal and glial cell determination. The protein encoded by this gene contains a conserved N-terminal GCM motif that has DNA-binding activity. The protein is a transcription factor that acts as a master regulator of parathyroid development. It has been suggested that this transcription factor might mediate the effect of calcium on parathyroid hormone expression and secretion in parathyroid cells. Mutations in this gene are associated with hypoparathyroidism.

Specifications

Antigen GCM2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Gcm2
Gene Accession No. O75603
Gene Alias Chorion-specific transcription factor GCMb; GCM motif protein 2; Gcm1; Gcm1-rs2; GCM2; Gcm-2; GCMB; gcmB transcription factor; Gcm-rs1; glial cell missing-like protein; glial cells missing 2; Glial cells missing homolog 2; glial cells missing homolog 2 (Drosophila); glial cells missing homolog b; glial cells missing transcription factor 2; glial cells missing-like protein 2; glide/gcm protein homolog; hGCMb; LOC100358264; mero; merovingian; mGCMb; RP3-417M14.3; transcription factor glial cells missing-2
Gene Symbols Gcm2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GCM2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9247
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.