Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCN1L1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553848
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553848 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553848 Supplier Invitrogen™ Supplier No. PA553848
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EALVTDAGEV TEAGKAYVPP RVLQEALCVI SGVPGLKGDV TDTEQLAQEM LIISHHPSLV AVQSGLWPAL LARMKIDPEA FITRHLDQII PRMTTQSPLN QSSMNAMGSL SVLSPDRVLP QLISTITA Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 92%.

Acts as a positive activator of the EIF2AK4/GCN2 protein kinase activity in response to amino acid starvation. Forms a complex with EIF2AK4/GCN2 on translating ribosomes; during this process, GCN1 seems to act as a chaperone to facilitate delivery of uncharged tRNAs that enter the A site of ribosomes to the tRNA-binding domain of EIF2AK4/GCN2, and hence stimulating EIF2AK4/GCN2 kinase activity. Participates in the repression of global protein synthesis and in gene-specific mRNA translation activation, such as the transcriptional activator ATF4, by promoting the EIF2AK4/GCN2-mediated phosphorylation of eukaryotic translation initiation factor 2 (eIF-2-alpha/EIF2S1) on 'Ser-52', and hence allowing ATF4-mediated reprogramming of amino acid biosynthetic gene expression to alleviate nutrient depletion. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCN1L1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GCN1
Gene Accession No. Q92616
Gene Alias 4932409G22; AL022764; eIF-2-alpha kinase activator GCN1; G431004K08Rik; Gcn1; GCN1 (general control of amino-acid synthesis 1, yeast)-like 1; GCN1 eIF2 alpha kinase activator homolog; GCN1 eIF-2-alpha kinase activator homolog; GCN1 general control of amino-acid synthesis 1-like 1; GCN1 general control of amino-acid synthesis 1-like 1 (yeast); GCN1, eIF2 alpha kinase activator homolog; GCN1L; GCN1L1; GCN1-like protein 1; General control of amino-acid synthesis 1-like protein 1; hsGCN1; KIAA0219; mKIAA0219; peroxisome proliferator activated receptor interacting complex protein; PRIC295; translational activator GCN1
Gene Symbols GCN1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GCN1L1. Recombinant protein control fragment (Product #RP-92376).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10985
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.