Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCN1L1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501561129
Encompass_Preferred
Change view
Click to view available options
20 μL; Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
50-156-1129 20 μL; Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1129 Supplier Bethyl Laboratories, Inc Supplier No. A301843AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Thioredoxin-interacting protein belongs to the arrestin family. It acts as an oxidative stress mediator by inhibiting thioredoxin activity or by inhibiting thioredoxin activity or by limiting its bioavailability. It functions as a transcriptional repressor, possibly by acting as a bridge molecule between transcription factors and corepressor complexes, and over-expression will induce G0/G1 cell cycle arrest. It is required for the maturation of natural killer cells. Recently reported, TXNIP deficiency led to the impairment of NLRP3 inflammasome activation to innate immune responses and reduced secretion of interleukin IL-1beta.

Specifications

Antigen GCN1L1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene GCN1
Gene Accession No. E9PVA8, Q92616
Gene Alias GCN1 (general control of amino-acid synthesis 1, yeast)-like 1, GCN1 eIF2 alpha kinase activator homolog, GCN1 eIF-2-alpha kinase activator homolog, GCN1 general control of amino-acid synthesis 1-like 1, GCN1 general control of amino-acid synthesis 1-like 1 (yeast), GCN1L, GCN1L1, GCN1-like protein 1, general control of amino-acid synthesis 1-like protein 1, hsGCN1, KIAA0219, peroxisome proliferator activated receptor interacting complex protein, PRIC295, translational activator GCN1
Gene Symbols Gcn1, Gcn1l1
Host Species Rabbit
Immunogen Between 2621 and 2671
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10985, 231659, 690632
Target Species Human, Mouse, Rat
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.