Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gcn1l1 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13315R

Description
Thioredoxin-interacting protein belongs to the arrestin family. It acts as an oxidative stress mediator by inhibiting thioredoxin activity or by inhibiting thioredoxin activity or by limiting its bioavailability. It functions as a transcriptional repressor, possibly by acting as a bridge molecule between transcription factors and corepressor complexes, and over-expression will induce G0/G1 cell cycle arrest. It is required for the maturation of natural killer cells. Recently reported, TXNIP deficiency led to the impairment of NLRP3 inflammasome activation to innate immune responses and reduced secretion of interleukin IL-1beta.Specifications
Gcn1l1 | |
Polyclonal | |
Unconjugated | |
GCN1 | |
4932409G22; AL022764; eIF-2-alpha kinase activator GCN1; G431004K08Rik; GCN1; GCN1 (general control of amino-acid synthesis 1, yeast)-like 1; GCN1 eIF2 alpha kinase activator homolog; GCN1 eIF-2-alpha kinase activator homolog; GCN1 general control of amino-acid synthesis 1-like 1; GCN1 general control of amino-acid synthesis 1-like 1 (yeast); GCN1, eIF2 alpha kinase activator homolog; GCN1L; GCN1L1; GCN1-like protein 1; General control of amino-acid synthesis 1-like protein 1; HsGCN1; KIAA0219; mKIAA0219; peroxisome proliferator activated receptor interacting complex protein; PRIC295; Translational activator GCN1 | |
Rabbit | |
Protein A | |
RUO | |
10985 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q92616 | |
GCN1 | |
KLH conjugated synthetic peptide derived from human Gcn1l1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction