Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCN2 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1001593JF549
Description
GCN2 Polyclonal antibody specifically detects GCN2 in Human samples. It is validated for Western BlotSpecifications
GCN2 | |
Polyclonal | |
Western Blot | |
eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1 | |
The immunogen recognized by this antibody maps to a region between residues 1 and 50 of human GCN2 (eIF2AK4, eukaryotic translation initiation factor 2 alpha kinase 4) using the numbering given in entry NP_001013725.2 (GeneID 440275). | |
0.1 mL | |
Phospho Specific, Protein Kinase | |
440275 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction