Learn More
Invitrogen™ GCN2 Recombinant Rabbit Monoclonal Antibody (1G2Z4)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA535353
Description
Immunogen sequence: MAGGRGAPGR GRDEPPESYP QRQDHELQAL EAIYGADFQD LRPDACGPVK EPPEINLVLY PQGLTGEEVY VKVDLRVKCP PTYPDVVPEI ELKNAKGLSN ESVNLLKSRL EELAKKHCGE VMIFELAYHV QSFLSEHNKP PPKSFHEEML ERRAQEEQQR LLEAKRKEEQ EQREILHEIQ RRKEEIKEEK KRKEMAKQER LEIASLSNQD HTSKKDPGGH RTAAILHGGS PDFVGNGKHR ANSSGRSRRE RQYSVCNSED SPGSCEILYF NMGSPDQLMV HKGKCIGSDE QLGKLVYNAL.
EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.
Specifications
GCN2 | |
Recombinant Monoclonal | |
1.0 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
D4A7V9, Q9P2K8, Q9QZ05 | |
EIF2AK4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GCN2 (NP_0010137252). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1G2Z4 | |
Unconjugated | |
EIF2AK4 | |
2610011M03; EIF2AK4; eIF-2-alpha kinase GCN2; eukaryotic translation initiation factor 2 alpha kinase 4; eukaryotic translation initiation factor 2-alpha kinase 4; Gcn2; GCN2 eIF2alpha kinase; GCN2-like protein; general control nonderepressible 2; Kiaa1338; mGCN2; PVOD2 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
114859, 27103, 440275 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.