Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCN2, p Thr899 Antibody (SR1336), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32151350UL
Description
GCN2 Monoclonal antibody specifically detects GCN2 in Human samples. It is validated for Western BlotSpecifications
GCN2 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1336 | |
Western Blot 1:500-1:2000 | |
eukaryotic translation initiation factor 2 alpha kinase 4, eukaryotic translation initiation factor 2-alpha kinase 4, GCN2GCN2 eIF2alpha kinase, GCN2-like protein, KIAA1338EC 2.7.11.1 | |
A synthesized peptide derived from human phospho-GCN2 [Thr899] (Uniprot #: Q9P2K8) | |
50 μg | |
Phospho Specific, Protein Kinase | |
440275 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction