Learn More
GCN2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A300555A
Description
The recommended shelf life for this product is 1 year from date of receipt.
EIF2AK4 (GCN2) is a metabolic-stress sensing protein kinase that phosphorylates the alpha subunit of eukaryotic translation initiation factor-2 in response to low amino acid availability. It plays a role as an activator of the integrated stress response required for adapatation to amino acid starvation. Mutations in the gene lead to pulmonary vennocclusive disease 2.Specifications
| GCN2 | |
| Polyclonal | |
| Unconjugated | |
| EIF2AK4 | |
| Eukaryotic translation initiation factor 2-alpha kinase 4; GCN2-like protein; EIF2AK4; GCN2; KIAA1338; | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 440275 | |
| 4° C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 1 mg/ml | |
| phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
| Q9P2K8 | |
| EIF2AK4 | |
| Between 1 and 50 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.