Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCN4 Antibody (C11L34), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP281273AF488
Description
GCN4 Monoclonal specifically detects GCN4 in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GCN4 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate | |
GCN4 | |
Biotinylated GCN4-peptide (HLENEVARLKK). | |
0.1 mL | |
Primary | |
This antibody binds to the HLENEVARLKK peptide from the leucine-zipper domain of the S. cerevisiae GCN4 transcriptional activator with picomolar affinity. | |
Store at 4°C in the dark. | |
IgG1 λ |
Western Blot, Immunocytochemistry, Immunofluorescence | |
C11L34 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
AAS101, AAS3, Amino acid biosynthesis regulatory protein, amino acid starvation-responsive transcription factor GCN4, ARG9, General Control Protein 4 | |
Mouse | |
Protein A purified | |
RUO | |
856709 | |
Yeast | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction