Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCN4 Antibody (C11L34) - Chimeric - Azide and BSA Free, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281274SS
Description
GCN4 Monoclonal specifically detects GCN4 in Yeast samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Surface Plasmon Resonance.Specifications
| GCN4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Surface Plasmon Resonance | |
| P03069 | |
| GCN4 | |
| Biotinylated GCN4-peptide (HLENEVARLKK). | |
| 0.025 mg | |
| Primary | |
| This antibody binds to the HLENEVARLKK peptide from the leucine-zipper domain of the S. cerevisiae GCN4 transcriptional activator with picomolar affinity. | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG λ |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| C11L34 | |
| Unconjugated | |
| PBS | |
| AAS101, AAS3, Amino acid biosynthesis regulatory protein, amino acid starvation-responsive transcription factor GCN4, ARG9, General Control Protein 4 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 856709 | |
| Yeast | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction