Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCN4 Chimeric Recombinant Mouse Monoclonal Antibody (C11L34)
GREENER_CHOICE

Catalog No. PIMA547957
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA547957 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA547957 Supplier Invitrogen™ Supplier No. MA547957
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Specificity: This antibody binds to the HLENEVARLKK peptide from the leucine-zipper domain of the S. cerevisiae GCN4 transcriptional activator with picomolar affinity.

GCN4 is a transcription factor protein in yeast that plays a crucial role in the regulation of genes involved in amino acid biosynthesis, such as HIS4, HIS7, HIS3, ARG1, ARG3, ARG4, and ARG5,6. The protein contains a basic leucine zipper (bZIP) domain that enables it to bind to specific DNA sequences, thereby regulating the transcription of target genes. The protein is also involved in the regulation of other cellular processes, including stress response and nutrient signaling, such as GSH1, YAP1, and TRX2. Its structure is composed of a dimeric coiled-coil domain, which allows for protein-protein interactions, and a DNA-binding bZIP domain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCN4 Chimeric
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry, Surface Plasmon Resonance
Classification Recombinant Monoclonal
Clone C11L34
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene GCN4
Gene Accession No. P03069
Gene Symbols GCN4
Host Species Mouse
Immunogen Biotinylated GCN4-peptide (HLENEVARLKK).
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 856709
Target Species Yeast
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.