Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCNF Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534020
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534020 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534020 Supplier Invitrogen™ Supplier No. PA534020
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GCNF Polyclonal Antibody for IHC (P)

This gene encodes an orphan nuclear receptor which is a member of the nuclear hormone receptor family. Its expression pattern suggests that it may be involved in neurogenesis and germ cell development. The protein can homodimerize and bind DNA, but in vivo targets have not been identified. The gene expresses at least alternatively spliced transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCNF
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene NR6A1
Gene Accession No. A0P8Z4, Q15406, Q64249
Gene Alias 1700113M01Rik; CT150; Gcnf; GCNF1; Germ cell nuclear factor; germ cell nuclear factor variant 1; h hRTR; hGCNF; hRTR; mGCNF; NCNF; NR61; NR6A1; Nuclear receptor subfamily 6 group A member 1; nuclear receptor subfamily 6, group A, member 1; retinoic acid receptor-related testis-associated receptor; retinoid receptor-related testis-specific receptor; RTR
Gene Symbols NR6A1
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from ligand-binding domain of human NR6A1 / GCNF.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100038028, 100070969, 100347187, 100773066, 14536, 2649, 362125, 541257, 609642
Target Species Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Hamster, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.