Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCNF Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565100
Description
Immunogen sequence: LILLSSLTVYS KQIFGELADV TAKYSPSDEE LHRFSDEGME VIERLIYLYH KFHQLKVSNE EYACMKAINF LNQDIRGLTS ASQLEQLNKR YWYICQDFTE Highest antigen sequence identity to the following orthologs - mouse 100%, rat 99%.
This gene encodes an orphan nuclear receptor which is a member of the nuclear hormone receptor family. Its expression pattern suggests that it may be involved in neurogenesis and germ cell development. The protein can homodimerize and bind DNA, but in vivo targets have not been identified. The gene expresses at least alternatively spliced transcript variants.
Specifications
GCNF | |
Polyclonal | |
Unconjugated | |
NR6A1 | |
1700113M01Rik; CT150; Gcnf; GCNF1; Germ cell nuclear factor; germ cell nuclear factor variant 1; h hRTR; hGCNF; hRTR; mGCNF; NCNF; NR61; NR6A1; Nuclear receptor subfamily 6 group A member 1; nuclear receptor subfamily 6, group A, member 1; retinoic acid receptor-related testis-associated receptor; retinoid receptor-related testis-specific receptor; RTR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2649 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15406 | |
NR6A1 | |
Recombinant Human GCNF. Recombinant protein control fragment (Product #RP-104633). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction