Learn More
GCNF Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 127121AP150UL

Description
This antibody specifically reacts with the 55 kDa NR6A1 protein.
This gene encodes an orphan nuclear receptor which is a member of the nuclear hormone receptor family. Its expression pattern suggests that it may be involved in neurogenesis and germ cell development. The protein can homodimerize and bind DNA, but in vivo targets have not been identified. The gene expresses at least alternatively spliced transcript variants.Specifications
GCNF | |
Polyclonal | |
Unconjugated | |
NR6A1 | |
GCNF, GCNF1, Germ cell nuclear factor, hGCNF, hRTR, NR61, NR6A1, RTR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14536, 2649, 362125 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15406, Q64249 | |
Nr6a1 | |
GCNF Fusion Protein Ag3395 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.