Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCNT1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA512146
Description
Glycosylation is one of the most universal but at the same time complex protein modifications. Modification with sugar moeties can be both co- translational and post- translational, occurring in the endoplasmatic reticulum and golgi. Three different forms of glycosylation can be distinguished: N-linked oligosaccharides, O-linked oligosaccharides and glycosyl- phosphatidylinositol (GPI-) anchors. Glycosylation results in thousands of distinct, bioactive glycoproteins resident throughout the cell that strongly determine protein-protein, carbohydrate-protein, membrane, and adhesion properties. Diseases associated with glycosylation defects include Congenital disorders of glycosylation, (CDG), also known as carbohydrate deficient glycoprotein syndromes, and diseases associated with advanced aging.
Specifications
GCNT1 | |
Polyclonal | |
Unconjugated | |
GCNT1 | |
5630400D21Rik; B130048E03; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N--acetylglucosaminyltransferase; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase (Core 2 branching enzyme) (Core2-GlcNAc-transferase) (C2GNT); beta1,6 N-acetylglucosaminyltransferase; beta-1,6-N-acetylglucosaminyltransferase; C2 GlcNAcT; C2GNT; C2GNT1; C2GNT-L; core 2 beta1,6 N-acetylglucosaminyltransferase-I; core 2 beta-1,6-N-acetylglucosaminyltransferase I; core 2 branching enzyme; core 2 GlcNAc-T; core 2 GnT; core 2-branching enzyme; Core2-GlcNAc-transferase; enzymatic glycosylation-regulating; G6NT; Gcnt1; glucosaminyl (N-acetyl) transferase 1, core 2; glucosaminyl (N-acetyl) transferase 1, core 2 (beta-1,6-N-acetylglucosaminyltransferase); glucosaminyl transferase 1, core 2; IGnT; NACGT2; NAGCT2 | |
Rabbit | |
Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
RUO | |
2650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 0.09% sodium azide | |
Q02742 | |
GCNT1 | |
KLH conjugated synthetic peptide between 399-428 amino acids from the C-terminal region of human GCNT1 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction