Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCNT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA512145
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA512145 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA512145 Supplier Invitrogen™ Supplier No. PA512145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GCNT1 Polyclonal Antibody for Western Blot, IHC (P)

Glycosylation is one of the most universal but at the same time complex protein modifications. Modification with sugar moeties can be both co- translational and post- translational, occurring in the endoplasmatic reticulum and golgi. Three different forms of glycosylation can be distinguished: N-linked oligosaccharides, O-linked oligosaccharides and glycosyl- phosphatidylinositol (GPI-) anchors. Glycosylation results in thousands of distinct, bioactive glycoproteins resident throughout the cell that strongly determine protein-protein, carbohydrate-protein, membrane, and adhesion properties. Diseases associated with glycosylation defects include Congenital disorders of glycosylation, (CDG), also known as carbohydrate deficient glycoprotein syndromes, and diseases associated with advanced aging.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCNT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GCNT1
Gene Accession No. Q02742, Q09324
Gene Alias 5630400D21Rik; B130048E03; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N--acetylglucosaminyltransferase; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase (Core 2 branching enzyme) (Core2-GlcNAc-transferase) (C2GNT); beta1,6 N-acetylglucosaminyltransferase; beta-1,6-N-acetylglucosaminyltransferase; C2 GlcNAcT; C2GNT; C2GNT1; C2GNT-L; core 2 beta1,6 N-acetylglucosaminyltransferase-I; core 2 beta-1,6-N-acetylglucosaminyltransferase I; core 2 branching enzyme; core 2 GlcNAc-T; core 2 GnT; core 2-branching enzyme; Core2-GlcNAc-transferase; enzymatic glycosylation-regulating; G6NT; Gcnt1; glucosaminyl (N-acetyl) transferase 1, core 2; glucosaminyl (N-acetyl) transferase 1, core 2 (beta-1,6-N-acetylglucosaminyltransferase); glucosaminyl transferase 1, core 2; IGnT; NACGT2; NAGCT2
Gene Symbols GCNT1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 88-117 amino acids from the central region of human GCNT1.
Purification Method Ammonium Sulfate Precipitation, Size-exclusion - Dialysis
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14537, 2650
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.