Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCNT2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | GCNT2 |
---|---|
Dilution | Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
GCNT2 Polyclonal specifically detects GCNT2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GCNT2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
2651 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:EVPWKYVINTCGQDFPLKTNREIVQHLKGFKGKNITPGVLPPDHAIKRTKYVHQEHTDKGGFFVKNTNILKTSPPHQLTIYFGTAYVALTREFVDFVLRDQRAIDLLQWSKDT | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Human | |
bA360O19.2, bA421M1.1, beta-1,6-N-acetylglucosaminyltransferase 2, cataract, congenital, CCAT, EC 2.4.1.150, GCNT2C, GCNT5, glucosaminyl (N-acetyl) transferase 2, I-branching enzyme, glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group), glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (Ii blood group), I beta-1,6-N-acetylglucosaminyltransferase, I-branching beta-1,6-acetylglucosaminyltransferase, I-branching enzyme, IGNTglucosaminyl (N-acetyl) transferase 5, II, Ii blood group, MGC163396, N-acetylglucosaminyltransferase, N-acetyllactosaminide beta-1,6-N-acetylglucosaminyltransferase, N-acetyllactosaminide beta-1,6-N-acetylglucosaminyl-transferase, NACGT1, NAGCT1, ULG3 | |
GCNT2 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title