Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCNT2 Polyclonal Antibody, PE, Bioss
SDP

Catalog No. 502281618
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1618 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1618 Supplier Bioss Supplier No. BS13316RPE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

GCNT2 Polyclonal antibody specifically detects GCNT2 in Human, Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen GCNT2
Applications Flow Cytometry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate PE
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Gcnt2
Gene Accession No. P97402, Q8N0V5
Gene Alias 5330430K10Rik; bA360O19.2; bA421M1.1; beta-1,6-N-acetylglucosaminyltransferase; beta-1,6-N-acetylglucosaminyltransferase 2; CCAT; CTRCT13; Gcnt2; GCNT2C; GCNT5; glucosaminyl (N-acetyl) transferase 2, I-branching enzyme; glucosaminyl (N-acetyl) transferase 2, I-branching enzyme (I blood group); glucosaminyltransferase, I-branching enzyme; I beta-1,6-N-acetylglucosaminyltransferase; I-branching beta-1,6-acetylglucosaminyltransferase; I-branching enzyme; IGNT; IGnTA; IGnTB; IGnTC; II; Ii blood group; large I antigen-forming beta-1,6-N-acetylglucosaminyltransferase; N-acetylglucosaminyltransferase; N-acetyllactosaminide beta-1,6-N-acetylglucosaminyl-transferase; NACGT1; NAGCT1; ULG3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human GCNT2
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14538, 2651
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.