Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCNT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532775
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532775 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532775 Supplier Invitrogen™ Supplier No. PA532775
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Dog, Panda, Horse (100%) Marmoset, Sheep, Bovine, Pig (93%).

This gene encodes a member of the N-acetylglucosaminyltransferase family. The encoded protein is a beta-6-N-acetylglucosamine-transferase that catalyzes the formation of core 2 and core 4 O-glycans on mucin-type glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCNT3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GCNT3
Gene Accession No. O95395
Gene Alias 2010013H22Rik; 2210021I22Rik; 2210401J11Rik; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3; beta1,6GlcNAc-transferase; beta-1,6-N-acetylglucosaminyltransferase; beta1,6-N-acetylglucosaminyltransferase-M; beta-16-N-acetylglucosaminyltransferase; C2/4GnT; C24GNT; C2GNT2; C2GNTM; C2GnT-M; C2GnT-mucin type; core 2 beta-1,6-N-acetylglucosaminyltransferase II; core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; dI/C2/C4GnT; dIGnT; Gcnt3; glucosaminyl (N-acetyl) transferase 3, mucin type; GNTM; GnT-M; hC2GnT-M; mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase
Gene Symbols GCNT3
Host Species Rabbit
Immunogen Synthetic 15 amino acid peptide from internal region of human GCNT3.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100068109, 487578, 9245
Target Species Human, Canine, Horse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.