Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCNT3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582417
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582417 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582417 Supplier Invitrogen™ Supplier No. PA582417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RLKCDSDHLG LESRESQSQY CRNILYNFLK LPAKRSINCS GVTRGDQEAV LQAILNNLEV KKKREPFTDT HYLSLTRDCE HFKAERKFIQ FPLSKEEVEF PIAYSMVIHE KIENFER Highest antigen sequence indentity to the following orthologs: Mouse - 72%, Rat - 73%.

This gene encodes a member of the N-acetylglucosaminyltransferase family. The encoded protein is a beta-6-N-acetylglucosamine-transferase that catalyzes the formation of core 2 and core 4 O-glycans on mucin-type glycoproteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCNT3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GCNT3
Gene Accession No. O95395
Gene Alias 2010013H22Rik; 2210021I22Rik; 2210401J11Rik; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3; beta1,6GlcNAc-transferase; beta-1,6-N-acetylglucosaminyltransferase; beta1,6-N-acetylglucosaminyltransferase-M; beta-16-N-acetylglucosaminyltransferase; C2/4GnT; C24GNT; C2GNT2; C2GNTM; C2GnT-M; C2GnT-mucin type; core 2 beta-1,6-N-acetylglucosaminyltransferase II; core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; dI/C2/C4GnT; dIGnT; Gcnt3; glucosaminyl (N-acetyl) transferase 3, mucin type; GNTM; GnT-M; hC2GnT-M; mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase
Gene Symbols GCNT3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GCNT3. Recombinant protein control fragment (Product #RP-90277).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9245
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.