Learn More
Invitrogen™ GCNT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589079
Description
Immunogen sequence: LKLSFRLKCD SDHLGLESRE SQSQYCRNIL YNFLKLPAKR SINCSGVTRG DQEAVLQAIL NNLEVKKKRE PFTDTHYLSL TRDCEHFKAE RKFIQFPLSK EEVEFPIAYS MVIHEKIENF ERLLRAVYAP QNIYCVHVDE KSPETFKEAV KAII; Positive Samples: 293T, SGC-7901, U-87MG; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
This gene encodes a member of the N-acetylglucosaminyltransferase family. The encoded protein is a beta-6-N-acetylglucosamine-transferase that catalyzes the formation of core 2 and core 4 O-glycans on mucin-type glycoproteins.
Specifications
GCNT3 | |
Polyclonal | |
Unconjugated | |
GCNT3 | |
2010013H22Rik; 2210021I22Rik; 2210401J11Rik; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 3; beta1,6GlcNAc-transferase; beta-1,6-N-acetylglucosaminyltransferase; beta1,6-N-acetylglucosaminyltransferase-M; beta-16-N-acetylglucosaminyltransferase; C2/4GnT; C24GNT; C2GNT2; C2GNTM; C2GnT-M; C2GnT-mucin type; core 2 beta-1,6-N-acetylglucosaminyltransferase II; core 2/core 4 beta-1,6-N-acetylglucosaminyltransferase; dI/C2/C4GnT; dIGnT; Gcnt3; glucosaminyl (N-acetyl) transferase 3, mucin type; GNTM; GnT-M; hC2GnT-M; mucus-type core 2 beta-1,6-N-acetylglucosaminyltransferase | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9245 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.19 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
O95395 | |
GCNT3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 27-180 of human GCNT3 (NP_0047421). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.