Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCNT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA568108
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA568108 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA568108 Supplier Invitrogen™ Supplier No. PA568108
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GCNT4 Polyclonal Antibody for Western Blot

Glycosyltransferase that mediates core 2 O-glycan branching, an important step in mucin-type biosynthesis. Does not have core 4 O-glycan or I-branching enzyme activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCNT4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GCNT4
Gene Accession No. Q9P109
Gene Alias 6-N-acetylg; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 4; C2GNT3; core 2 beta-1,6-N-acetylglucosaminyltransferase 3; core 2-branching enzyme 3; Core2-GlcNAc-transferase 3; Gcnt4; glucosaminyl (N-acetyl) transferase 3, mucin type pseudogene; glucosaminyl (N-acetyl) transferase 4; glucosaminyl (N-acetyl) transferase 4, core 2; glucosaminyl (N-acetyl) transferase 4, core 2 (beta-1,6-N-acetylglucosaminyltransferase); glucosaminyl transferase 4, core 2; Gm279; Gm73
Gene Symbols GCNT4
Host Species Rabbit
Immunogen Synthesized peptide derived from human GCNT4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51301
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.