Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCNT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101454
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101454 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101454 Supplier Invitrogen™ Supplier No. PA5101454
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GCNT6.

GCNT2P1 (GCNT2 Pseudogene 1) is a Pseudogene. Among its related pathways are O-linked glycosylation and Metabolism of proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCNT6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GCNT2P1
Gene Accession No. 0
Gene Symbols GCNT2P1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GCNT6, corresponding to a region within N-terminal amino acids.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644378
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.