Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCNT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562651
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562651 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562651 Supplier Invitrogen™ Supplier No. PA562651
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TNREIIHYIR SKWSDKNITP GVIQPLHIKS KTSQSHLEFV PKGSIYAPPN NRFKDKPP Highest antigen sequence identity to the following orthologs: Mouse - 48%, Rat - 55%.

GCNT7, is a 430 amino acid glycosyltransferase that is localized to the Golgi apparatus. Other members of this family include GCNT1, GCNT2, GCNT3, GCNT4 and GCNT6. GCNT1 has been shown to play an important regulatory role in the biosynthesis of mucin-type O-glycans, which serve as ligands in cell adhesion. Specifically, GCNT1 expression in leukocytes regulates the synthesis of core 2 O-glycans that carry sialyl-Lewis x (sLex) oligosaccharides, which confer high affinity binding to Selectin proteins. Since downregulation of Selectin ligand expression has been shown to inhibit tissue infiltration, glycosyltransferase 14 family members represent potential drug targets for the treatment of inflammatory disorders and other pathologies involving Selectin proteins.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCNT7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GCNT7
Gene Accession No. Q6ZNI0
Gene Alias A330041C17Rik; beta 1,6-N-acetylglucosaminyltransferase; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 7; beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,6-N-acetylglucosaminyltransferase 7-like; C20orf105; dJ1153D9.2; gcnt; GCNT7; glucosaminyl (N-acetyl) transferase family member 7; gnt; likely orthologue of H. sapiens chromosome 20 open reading frame 105 (C20orf105); LOC102550196
Gene Symbols GCNT7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GCNT7. Recombinant protein control fragment (Product #RP-99334).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140687
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.