Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCOM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
| Antigen | GCOM1 |
|---|---|
| Concentration | 0.43 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2002
![]()
|
Proteintech Group Inc
181291AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
GCOM1, also named as Myozap, Gup and Gcom, belongs to the GCOM1 family. It plays a role in cellular signaling via Rho-related GTP-binding proteins and subsequent activation of transcription factor SRF. GCOM1 has 11 isoforms with MW 44-54 kDa and 64 kDa.Specifications
| GCOM1 | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q3UIJ9, Q5EB94 | |
| 102371, 145781, 363091 | |
| GCOM1 Fusion Protein Ag12708 | |
| Primary | |
| -20°C | |
| GCOM1 |
| 0.43 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Gcom, GCOM1, Gcom2, GRINL1A, GRINL1A combined protein, GRINL1A complex locus, GRINL1A upstream protein, Gup, Gup1, Gup2, MYOZAP, MYZAP, POLR2M | |
| GCOM1, Myzap | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title