Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GCP2, Mouse anti-Mouse, Chicken, Human, Frog, Fish, Clone: GCP2-02, Millipore Sigma™

Catalog No. MABT1322100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT1322100 100 μg
MABT132225 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABT1322100 Supplier MilliporeSigma Supplier No. MABT1322100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-GCP2, clone GCP2-02, Cat. No. MABT1322, is a mouse monoclonal antibody that detects Gamma-tubulin complex component 2 (CGP2) and has been tested for use in ELISA, Electron Microscopy, Immunocytochemistry, Immunoprecipitation, and Western Blotting.

Gamma-tubulin complex component 2 (UniProt: Q921G8; also known as GCP2) is encoded by the Tubgcp2 (also known as Gcp2) gene (Gene ID: 74237) in murine species. Gamma-tubulin complex that is essential for microtubule nucleation at the centrosome is composed of gamma-tubulin, GCP2, GCP3, GCP4, GCP5 and GCP6. GCPs are characterized by sequence homology in two specific regions known as the grip1 and grip2 motifs. GPCs associate with gamma-tubulin to form a V-shaped subcomplex known as gamma-tubulin small complex. In this complex GCPs 2 and 3 interact via their N-terminal domain and form the arms of the V. The two-distal part of the V (C-terminal domains) bind to gamma-tubulin. The gamma-tubulin complex serves as a template for polarized growth and regulation of microtubules in eukaryotes. Significantly higher expression of both GPC2 and 3 have been reported in glioblastoma cells and they are concentrated I the centrosomes in interphase glioblastoma cells. Depletion of GPC2 and 3 causes accumulation of cells in G2/M and result in mitotic delay. (Ref.: Farache, D., et al. (2016). J. Biol. Chem. 291(44); 23112-25; Draberova, E., et al. (2015). J. Neuropathol. Exp. Neurol. 74(7); 723-42).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCP2
Applications ELISA, Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone GCP2-02
Conjugate Unconjugated
Dilution Western Blotting Analysis: 1μg/mL from a representative lot detected GCP2 in human glioblastoma T98G cell lysate.Immunoprecipitation Analysis: A representative lot immunoprecipitated GCP2 in Immunoprecipitation applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).Immunocytochemistry Analysis: A representative lot detected GCP2 in Immunocytochemistry applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).Western Blotting Analysis: A representative lot detected GCP2 in Western Blotting applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).ELISA Analysis: A representative lot detected GCP2 in ELISA applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42).Electron Microscopy Analysis: A representative lot detected GCP2 in Electron Microscopy applications (Draberova, E., et. al. (2015). J Neuropathol Exp Neurol. 74(7):723-42). Immunoprecipitation Analysis: A represnetative lot immunoprecipitated GCP2 in mouse bone-marrow mast cells (Courtesy of Pavel Draber, Ph.D., Institute of Molecular Genetics of the ASCR, v.v.i., Prague, Czech Republic).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Gamma-tubulin complex component 2
Gene Symbols Tubgcp2;Gcp2
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to the first 194 amino acids from the N-terminal region of murine Gamma-tubulin complex component 2.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Gene ID (Entrez) NP_001272936.1
Test Specificity Clone GCP2-02 specifically detects GCP2 in T98G glioblastoma cells. It targets an epitope within 12 amino acids from the N-terminal region.
Target Species Mouse, Chicken, Human, Frog, Fish
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.