Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA575797
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
The γ-Tubulin complex is composed of γ Tubulin and the γ-Tubulin complexassociated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. γ-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the γ-Tubulin complex. GCP2 (γ-Tubulin complex component 2), also known as TUBGCP2 or SPBC97 (spindle pole body protein Spc97 homolog) is a ubiquitously expressed 902 amino acid protein that localizes to the centrosome and is involved in microtubule nucleation.
Specifications
GCP2 | |
Polyclonal | |
Unconjugated | |
TUBGCP2 | |
1700022B05Rik; a disintegrin and metalloprotease domain 8; Adam8; Gamma-ring complex protein 103 kDa; gamma-tubulin complex component 2; GCP2; GCP-2; Grip103; h103p; hGCP2; hGrip103; hSpc97; SPBC97; Spc97p; spindle pole body protein Spc97 homolog; Tubgcp2; tubulin gamma complex associated protein 2; tubulin, gamma complex associated protein 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10844, 309098, 74237 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q921G8, Q9BSJ2 | |
TUBGCP2 | |
Synthetic peptide corresponding to residues in Human TUBGCP2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction