Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560549
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560549 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560549 Supplier Invitrogen™ Supplier No. PA560549
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLSVLHSQLQ LEDDQGVNLG LESSLTLRRL LVWTYDPKIR LKTLAALVDH CQGRKGGELA SAVHAYTKTG DPYMRSLVQH ILSLVSHPVL SFLYRWIYDG ELEDTYHEFF VASDPTVKT Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 98%.

The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complex associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP3 (gamma-tubulin complex component 3), also known as TUBGCP3 or SPBC98, localizes to the centrosome and is an ubiquitously expressed 907 amino acid member of the gamma-Tubulin complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TUBGCP3
Gene Accession No. Q96CW5
Gene Alias 104p; gamma-ring complex protein 104 kDa; Gamma-tubulin complex component 3; GCP3; GCP-3; Grip104; h104p; hGCP3; hGrip104; hSpc98; SPBC98; Spc98; Spc98p; spindle pole body protein; spindle pole body protein Spc98 homolog; Tubgcp3; tubulin gamma complex associated protein 3; tubulin, gamma complex associated protein 3
Gene Symbols TUBGCP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GCP3. Recombinant protein control fragment (Product #RP-97940).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10426
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.