Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GCP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA537451
Description
This antibody detects endogenous protein at a molecular weight of 104 kDa. Purity is >95% by SDS-PAGE.
The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complex associated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP3 (gamma-tubulin complex component 3), also known as TUBGCP3 or SPBC98, localizes to the centrosome and is an ubiquitously expressed 907 amino acid member of the gamma-Tubulin complex.
Specifications
GCP3 | |
Polyclonal | |
Unconjugated | |
TUBGCP3 | |
104p; gamma-ring complex protein 104 kDa; Gamma-tubulin complex component 3; GCP3; GCP-3; Grip104; h104p; hGCP3; hGrip104; hSpc98; SPBC98; Spc98; Spc98p; spindle pole body protein; spindle pole body protein Spc98 homolog; Tubgcp3; tubulin gamma complex associated protein 3; tubulin, gamma complex associated protein 3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10426, 259279, 306599 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P58854, Q96CW5 | |
TUBGCP3 | |
Synthetic peptide corresponding to amino acids 204-252 of human GCP3. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction