Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101601
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101601 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101601 Supplier Invitrogen™ Supplier No. PA5101601
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TUBGCP4.

The gamma-Tubulin complex is composed of gamma Tubulin and the gamma-Tubulin complexassociated proteins GCP2, GCP3, GCP4, GCP5 and GCP6, all of which are essential components of microtubule organizing centers. gamma-Tubulin complex components are localized to both the centrosome, where they are involved in microtubule nucleation, and to the cytoplasm, where they exist as soluble complexes that can be recruited to the centrosome as needed. Although the GCP proteins are related, they have distinct roles which contribute to the proper function of the gamma-Tubulin complex. GCP4 (gamma-Tubulin complex component 4), also known as TUBGCP4, is a ubiquitously expressed 667 amino acid member of the gamma-Tubulin complex that localizes to the metaphase spindle during mitosis. In response to proteosome inhibition, GCP4 exhibits increased accumulation at the pericentiolar material where it participates in microtubule organization and nucleation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCP4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TUBGCP4
Gene Accession No. Q9D4F8, Q9UGJ1
Gene Alias 4932441P04Rik; 76P; D2Ertd435e; gamma tubulin ring complex protein; gamma tubulin ring complex protein (76p gene); gamma-ring complex protein 76 kDa; Gamma-tubulin complex component 4; GCP4; GCP-4; Grip76; h76p; hGCP4; hGrip76; MCCRP3; RGD1306924; TUBGCP4; tubulin gamma complex associated protein 4; tubulin, gamma complex associated protein 4
Gene Symbols TUBGCP4
Host Species Rabbit
Immunogen A synthesized peptide derived from human TUBGCP4(Accession Q9UGJ1), corresponding to amino acid residues H193-L243.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27229, 51885
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.