Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCR2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299443MFV610
Description
GCR2 Polyclonal antibody specifically detects GCR2 in Human, Mouse, Cynomolgus Monkey samples. It is validated for Immunohistochemistry (Paraffin)Specifications
GCR2 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ecdysoneless homolog (Drosophila), GCR2, HSGT1, Protein ecdysoneless homolog, protein SGT1, SGT1, Suppressor of GCR2, suppressor of S. cerevisiae gcr2, X | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human GCR2 | |
0.1 mL | |
Cell Cycle and Replication | |
11319 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Cynomolgus Monkey | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction