Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GCR2 Antibody (PCRP-ECD-1D10), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314278FR
This item is not returnable.
View return policy
Description
GCR2 Monoclonal antibody specifically detects GCR2 in Human samples. It is validated for Flow Cytometry, ImmunofluorescenceSpecifications
GCR2 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunofluorescence | |
PCRP-ECD-1D10 | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
ecdysoneless homolog (Drosophila), GCR2, HSGT1, Protein ecdysoneless homolog, protein SGT1, SGT1, Suppressor of GCR2, suppressor of S. cerevisiae gcr2, X | |
Recombinant full-length human GCR2 protein (Uniprot: O95905) | |
0.1 mL | |
Cell Cycle and Replication | |
11319 | |
Store at 4°C in the dark. | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction