Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCSAML Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA546340
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA546340 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA546340 Supplier Invitrogen™ Supplier No. PA546340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: DEERKRQEMT TFERKLQDQD KKSQEVSSTS NQENENGSGS EEVCYTVINH Sequence homology: Dog: 100%; Human: 100%.

This gene encodes a protein thought to be a signaling molecule associated with germinal centers, the sites of proliferation and differentiation of mature B lymphocytes. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GCSAML
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GCSAML
Gene Accession No. Q5JQS6
Gene Alias C1orf150; GCSAML; germinal center associated signaling and motility like; germinal center-associated signaling and motility-like protein; germinal center-associated, signaling and motility-like
Gene Symbols GCSAML
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GCSAML (aa 25-74).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 148823
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.