Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GCSAML Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA555639

Catalog No. PIPA555639


Only null left
Add to Cart

Description

Description

Immunogen sequence: MGNYLLRKLS CLGENQKKPK KGNPDEERKR QEMTTFERKL QDQDKKSQEV SSTSNQENEN GSGSEEV Highest antigen sequence identity to the following orthologs: Mouse - 30%, Rat - 30%.

This gene encodes a protein thought to be a signaling molecule associated with germinal centers, the sites of proliferation and differentiation of mature B lymphocytes. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GCSAML
Polyclonal
Unconjugated
GCSAML
C1orf150; GCSAML; germinal center associated signaling and motility like; germinal center-associated signaling and motility-like protein; germinal center-associated, signaling and motility-like
Rabbit
Antigen Affinity Chromatography
RUO
148823
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.2 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5JQS6
GCSAML
Recombinant protein corresponding to Human GCSAML. Recombinant protein control fragment (Product #RP-94447).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.